Safety spectacles, i-works
Supplier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9194.171 9194.175 9194.270 9194.365
UVEX9194.171EA 29.4 GBP
UVEX9194.171 UVEX9194.175 UVEX9194.270 UVEX9194.365
Safety spectacles, i-works
Safety Glasses
The 100% metal free uvex i-works encapsulates the best of uvex eyewear: the sporty design, panoramic lens and low weight make it extremely comfortable and together with the unrivalled permanent uvex lens coating technology (uvex supravision excellence), provides the highest level of optical clarity remaining fog free even after repeated cleaning. High-tech soft grip side arm materials further enhance comfort levels delivering a stable, non-slip fit.
  • Sporty design and panoramic lens for great lateral vision
  • High performance anti-fog coating on the inside,
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable