Welding cover spectacles, uvex super f OTG
Supplier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9169.543 9169.545 9169.541
UVEX9169.543EA 55.5 GBP
UVEX9169.543 UVEX9169.545 UVEX9169.541
Welding cover spectacles, uvex super f OTG
Safety Glasses
The modern protective welding cover spectacles fit over most prescription spectacles.
  • New side arm hinge concept with extremely soft ear pieces
  • Permanent anti-fog coating on the inside of the lens
  • Perfect colour recognition in line with uvex sun glare filters
  • Outstanding protection against mechanical hazards
  • Minimum damage from welding sparks
  • UV and IR radiation protection