Safety spectacles, uvex winner
Supplier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9159.105 9159.005 9159.118
111-8215EA 64 GBP
111-8215 UVEX9159.005 UVEX9159.118
Safety spectacles, uvex winner
Safety Glasses
These metal dual lens spectacles provide clear visibility in all directions. The large lenses protect the whole eye area.
  • Close fitting offers high levels of protection and a wide field of visibility
  • Sprung side arms provide secure fit
  • Hinged nose pads adapt to the wearer
  • Comfortable fit, even when worn for extended periods
Certifications: EN 166, EN 168, EN 170, EN 172
Order Now
SPECIFICATIONS
Environmentally Preferable