Chemical resistant gloves, uvex profaviton BV-06
Supplier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6095708 6095709 6095710 6095711
UVEX6095708EA 282 GBP
UVEX6095708 UVEX6095709 UVEX6095710 UVEX6095711
Chemical resistant gloves, uvex profaviton BV-06
Gloves
These chemical protection gloves are made from butyl rubber with a Viton® outer layer.
  • Good grip in damp and wet areas
  • Good fit
  • High flexibility
  • Rolled edge
Ideal for the chemical industry as they provide good resistance to aliphatic and aromatic hydrocarbons (e.g. hexane, benzene, toluene, xylene and others), halogenated hydrocarbons (e.g. trichloroethylene, perchloroethylene, dichloromethane and others), organic and inorganic acids (diluted to concentrated), as well as saturated saline solutions.
Certifications: EN 388:2016 (2120A), EN ISO 374-1:2More Product Information