Disposable earplugs, uvex hi-com detec
Supplier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2112.114
UVEX2112.114EA 186 GBP
UVEX2112.114
Disposable earplugs, uvex hi-com detec
Earplugs Disposable Earplugs
The hi-com detec is a detectable disposable earplug for the food industry. The innovative shape of the hi-com reduces the level of sound that can build up in the inner ear when wearing hearing protection. There is no risk of overprotection in areas with medium noise exposure.
  • Earplug design make it possible to understand speech therefore improving communication
  • Easy to insert, even in narrower ear canals
  • Metal powder in cord allows full detection, even if the cord is shredded
  • Blue colour enables easy recognition in production areas
SNR: 24 dB
Certifications: EN 352-2:2002