Anti-Thyroid Iodide Transporter Rabbit Polyclonal Antibody

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

T5610-01-100
USBIT5610-01-100EA 696 GBP
USBIT5610-01-100
Anti-Thyroid Iodide Transporter Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Thyroid Iodide Transporter
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR