Anti-Stat 6 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

S7972-40M-FITC
USBIS7972-40M-FITCEA 829 GBP
USBIS7972-40M-FITC
Anti-Stat 6 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antibodies
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: Stat 6
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR