Anti-STAT3 Rabbit Polyclonal Antibody

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

S7971-20G
USBIS7971-20GEA 476 GBP
USBIS7971-20G
Anti-STAT3 Rabbit Polyclonal Antibody
Antibodies
  • Recommended dilution: ELISA:1:2500; Western Blot:1:2500; Immunochemistry:1:100

Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR