Anti-Pantothenate Kinase 2, Mitochondrial Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P3048-71-BIOT
USBIP3048-71-BIOTEA 829 GBP
USBIP3048-71-BIOT
Anti-Pantothenate Kinase 2, Mitochondrial Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: Pantothenate Kinase 2, Mitochondrial
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR