Anti-Mycoplasma pneumoniae Mouse Monoclonal Antibody [clone: 9H530]

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M9750-15H
USBIM9750-15HEA 617 GBP
USBIM9750-15H
Anti-Mycoplasma pneumoniae Mouse Monoclonal Antibody [clone: 9H530]
Antibodies
Type: Primary
Antigen: Mycoplasma pneumoniae
Clonality: monoclonal
Clone: 9H530
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR