Anti-Mycoplasma hominis Mouse Monoclonal Antibody [clone: 9L503]

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M9750-06D
USBIM9750-06DEA 465 GBP
USBIM9750-06D
Anti-Mycoplasma hominis Mouse Monoclonal Antibody [clone: 9L503]
Antibodies
Type: Primary
Antigen: Mycoplasma hominis
Clonality: monoclonal
Clone: 9L503
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR