Middlebrook 7H10 OADC (Oleic Albumin Dextrose Catalase) Growth Supplement
Supplier: US Biological
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M3895-01-500G
USBIM3895-01-500GEA 889 GBP
USBIM3895-01-500G
Middlebrook 7H10 OADC (Oleic Albumin Dextrose Catalase) Growth Supplement
Cell Culture Supplements and Additives
Enrichment supplement recommended for the cultivation of Mycobacteria sp. OADC enrichment medium is used with Middlebrook 7H10 Mycobacteria Agar (Powder).
  • Light beige with slight green tint
  • Homogeneous
  • Free flowing powder
Solubility: Light yellow, turbid.