Anti-IgG Rabbit Polyclonal Antibody (Texas Red®)

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

I1904-63E
USBII1904-63EEA 575 GBP
USBII1904-63E
Anti-IgG Rabbit Polyclonal Antibody (Texas Red®)
Antibodies
Type: Secondary
Antigen: IgG (IgG)
Clonality: polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Rabbit
Isotype:
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR