Anti-Glycerol-3-Phosphate Dehydrogenase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

G8154-72N-HRP
USBIG8154-72N-HRPEA 829 GBP
USBIG8154-72N-HRP
Anti-Glycerol-3-Phosphate Dehydrogenase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: Glycerol-3-Phosphate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol GPDH
Environmentally Preferable
Antigen name Glycerol-3-Phosphate Dehydrogenase
Antibody type Primary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitope N-terminal
Form liquid
Purification purified by immunoaffinity chromatography
Storage buffer PBS, pH 7,2
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR