Anti-GluR1,2 delta Rabbit Polyclonal Antibody

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

G3500-10A
USBIG3500-10AEA 707 GBP
USBIG3500-10A
Anti-GluR1,2 delta Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:5000

Type: Primary
Antigen: GluR1,2 delta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR