Anti-Cytosine, 5-Methyl Sheep polyclonal antibody

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C9111
USBIC9111EA 360 GBP
USBIC9111
Anti-Cytosine, 5-Methyl Sheep polyclonal antibody
Antibodies
Recommended dilution: Western Blot: 1:500; Immunoprecipitation: 25-100 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: Cytosine, 5-Methyl
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Order Now

SPECIFICATIONS

Antigen name Cytosine, 5-Methyl
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity
Host Sheep
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Form liquid
Purification purified by Protein G chromatography
Concentration ~1 mg/ml
Storage buffer PBS, 40% glycerol
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR