Anti-KRT16 Rabbit Polyclonal Antibody

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C9097-15P4
USBIC9097-15P4EA 632 GBP
USBIC9097-15P4
Anti-KRT16 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Immunochemistry: 1:25-200

Type: Primary
Antigen: KRT16 (keratin 16)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR