Anti-Cysteine-rich, Angiogenic Inducer 61 Mouse Monoclonal Antibody [clone: 10C314]

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C9018-01E
USBIC9018-01EEA 479 GBP
USBIC9018-01E
Anti-Cysteine-rich, Angiogenic Inducer 61 Mouse Monoclonal Antibody [clone: 10C314]
Antibodies
Recommended dilution: Western Blot: 1:250-1:1000

Type: Primary
Antigen: Cysteine-rich, Angiogenic Inducer 61
Clonality: Monoclonal
Clone: 10C314
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR