Anti-CCND3 Mouse monoclonal antibody [clone: 11n57]

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C8454-25B
USBIC8454-25BEA 752 GBP
USBIC8454-25B
Anti-CCND3 Mouse monoclonal antibody [clone: 11n57]
Antibodies
Recommended dilution: Western Blot: 1 ?g/ml; Immunochemistry: 8-25 ?g/ml

Type: Primary
Antigen: CCND3 (cyclin D3)
Clonality: Monoclonal
Clone: 11n57
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR