Anti-Serum Albumin Goat Polyclonal Antibody

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A1327-02B
USBIA1327-02BEA 386 GBP
USBIA1327-02B
Anti-Serum Albumin Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Serum Albumin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol HSA
Environmentally Preferable
Antigen name Human Serum Albumin
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Goat
Isotype IgG
Format Serum
Form liquid
Storage buffer TBS, pH 7,4, 0,09% sodium azide.
Storage temperature Cool
Shipping temperature Ice
Tested applications Turbidimetry

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR