Anti-AIRE Goat Polyclonal Antibody

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A1059-88R
USBIA1059-88REA 639 GBP
USBIA1059-88R
Anti-AIRE Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:32000; Western Blot: 0.5-1 ?gml; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: AIRE (autoimmune regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR