Anti-OGFOD2 Rabbit Polyclonal Antibody

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

146802
USBI146802EA 607 GBP
USBI146802
Anti-OGFOD2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: OGFOD2 (2-oxoglutarate and iron-dependent oxygenase domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR