Anti-Scavenger Receptor BI Rabbit Polyclonal Antibody

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

146450
USBI146450EA 607 GBP
USBI146450
Anti-Scavenger Receptor BI Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1-2?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: Scavenger Receptor BI
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen name Scavenger Receptor BI
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Form liquid
Purification purified by immunoaffinity chromatography
Concentration ~1 mg/ml
Storage buffer PBS, 0,02% sodium azide.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR