Anti-STAR Mouse Monoclonal Antibody [clone: 5F9]

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

133924
USBI133924EA 614 GBP
USBI133924
Anti-STAR Mouse Monoclonal Antibody [clone: 5F9]
Antibodies
Type: Primary
Antigen: STAR (steroidogenic acute regulatory protein)
Clonality: monoclonal
Clone: 5F9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR