Anti-Elongin B Mouse Monoclonal Antibody [clone: 6F6]

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

126294
USBI126294EA 614 GBP
USBI126294
Anti-Elongin B Mouse Monoclonal Antibody [clone: 6F6]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: Elongin B
Clonality: monoclonal
Clone: 6F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Rat
Order Now

SPECIFICATIONS

Antigen symbol ELOB
Environmentally Preferable
Antigen name Elongin B
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 6F6
Reactivity Human,Rat
Host Mouse
Isotype IgG2a kappa
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Amino acid number 9 to 119
Sequence RHKTTIFTDAKESSTVFELKRIVEGILKRPPDEQRLYKDDQLLDDGKTLGECGFTSQTARPQAPATVGLAFRADDTFEALCIEPFSSPPELPDVMKPQDSGSSANEQAVQ
Form liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2.
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR