Anti-DBF4 Mouse Monoclonal Antibody [clone: 6G9]

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

125648
USBI125648EA 614 GBP
USBI125648
Anti-DBF4 Mouse Monoclonal Antibody [clone: 6G9]
Antibodies
Recommended dilution: Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: DBF4 (DBF4 zinc finger)
Clonality: monoclonal
Clone: 6G9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR