Anti-TNKS Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

043098-HRP
USBI043098-HRPEA 829 GBP
USBI043098-HRP
Anti-TNKS Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: TNKS (tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol TNKS
Environmentally Preferable
Antigen name tankyrase, TRF1-interacting ankyrin-related ADP-ribose polymerase
Antibody type Primary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Epitope N-terminal
Amino acid number 54 to 82
Form liquid
Purification purified by Protein A chromatography
Storage buffer PBS, pH 7,2
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR