Anti-PEAK1 Rabbit Polyclonal Antibody

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

041661
USBI041661EA 626 GBP
USBI041661
Anti-PEAK1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: PEAK1 (pseudopodium-enriched atypical kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR