Anti-CHRNA3 Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

033871-BIOTIN
USBI033871-BIOTINEA 829 GBP
USBI033871-BIOTIN
Anti-CHRNA3 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: CHRNA3 (cholinergic receptor, nicotinic, alpha 3 (neuronal))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR