Anti-BOLL Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

032559-AP
USBI032559-APEA 829 GBP
USBI032559-AP
Anti-BOLL Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: BOLL (boule-like RNA-binding protein)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR