Anti-Rat Serum Albumin Rabbit Polyclonal Antibody

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

029012
USBI029012EA 292 GBP
USBI029012
Anti-Rat Serum Albumin Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Rat Serum Albumin (Rat Serum Albumin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR