Anti-Pigeon Serum Albumin Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

029005
USBI029005EA 474 GBP
USBI029005
Anti-Pigeon Serum Albumin Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: Pigeon Serum Albumin (Pigeon Serum Albumin)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR