Anti-Guinea Pig Serum Albumin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

028998
USBI028998EA 474 GBP
USBI028998
Anti-Guinea Pig Serum Albumin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: Guinea Pig Serum Albumin (Guinea Pig Serum Albumin)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Guinea pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR