Anti-14-3-3 sigma Goat Polyclonal Antibody

Supplier: US Biological
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

0014-19C
USBI0014-19CEA 645 GBP
USBI0014-19C
Anti-14-3-3 sigma Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:64000; Western Blot: 0.1-0. 3?g/ml

Type: Primary
Antigen: 14-3-3 sigma (14-3-3 sigma)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR