Instrument trays
Supplier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4743 4744
USBE4743EA 74.6 GBP
USBE4743 USBE4744
Instrument trays
Trays/Dishes/Pots Instrument Trays/Dishes
Made of stainless steel.
  • Rounded corners
  • With rim
  • Very good chemical and temperature resistance