Spoon spatulas
Supplier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3346 3347 3348
USBE3346EA 6.2 GBP
USBE3346 USBE3347 USBE3348
Spoon spatulas
Spatulas
Stainless steel.
  • Double-ended: Spatula and spoon end
  • Spoon width: 29 mm