Cover spectacles, x-Generation, 5X7
Supplier: UNIVET
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5X7.01.11.30 5X7.03.11.00 5X7.01.11.02 5X7.01.00.00
UNVT5X7.01.11.30EA 266 GBP
UNVT5X7.01.11.30 UNVT5X7.03.11.00 UNVT5X7.01.11.02 UNVT5X7.01.00.00
Cover spectacles, x-Generation, 5X7
Safety Glasses
The model 5X7 is the ultimate OTG: it provides a maximum of protection and the comfort of operating with prescription spectacles.
  • Ideal for use with prescription spectacles
  • Frame design prevents the overlapping of the temples
  • Adjustment of temples in length with SoftPad technology
  • Wide range of filters
Weight: 37 g