Hypodermic needles, MONOJECT™
Supplier: TYCO HEALTHCARE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8881200037
TYCO8881200037EA 0 GBP
TYCO8881200037
Hypodermic needles, MONOJECT™
Syringe Needles
The MONOJECT™ hypodermic needles are ultra-sharp, tri-beveled, anti-coring, hypodermic needles.
  • Latex free
  • Precision machined Luer lock aluminum hub
  • Colour coded needle sheath
  • Can be autoclaved or gas sterilised for use in O.R./procedure packs
A Bevel - Regular bevel for intramuscular and subcutaneous injections
B Bevel - Short bevel for intravenous or intra-arterial applications and nerve blocks