Control slide, Fungi TISSUE-TROL™, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TTR004-25EA
TTR004-25EAEA 231 GBP
TTR004-25EA
Control slide, Fungi TISSUE-TROL™, Sigma-Aldrich®
Slides Microscopy Control Slides
Histology slides with paraffin embedded section of mouse lung tissue containing Candida albicans.
Suitable for use with Silver Stain (Modified GMS) Procedure No. HT100.