Disinfectant, non sterile chlorine disinfectant, Zircon
Supplier: TRISTEL SOLUTIONS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CRY506
TRSTCRY506EA 0 GBP
TRSTCRY506
Disinfectant, non sterile chlorine disinfectant, Zircon
Cleaning Agents Disinfectants
A non foaming, sodium hypochlorite based, disinfectant.
  • Ideal for use on walls, floors and equipment
  • Suitable for use in CIP and spraywash enclosed systems
  • Dilution for use: 0,5 to 2% v/v
Areas of use include walls, floors and equipment such as CIP and spraywash enclosed systems. Zircon is not soft metal safe, but is suitable for use on stainless steel and is also ideal for drain and mop disinfection.
All disinfectants are supported by comprehensive technical files which provide all the documentation for validation. All disinfectants are supportedMore Product Information