Flasks, Ultra Yield™
Supplier: THOMSON INSTRUMENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
931136-B 931144 931141 931138
THMS931136-BEA 505 GBP
THMS931136-B THMS931144 THMS931141 THMS931138
Flasks, Ultra Yield™
Flasks Cell Culture Flasks
Thomson's Ultra Yield™ flasks are so named because these flasks actually enhance the growth of E.coli as well as other microbial cells.
The AirOtop® Seals are designed as a single-use, disposable seal. Enhanced AirOtop® Seals offer simple, disposable, sterile high air exchange. Seals fit all shake flasks. The Enhanced AirOtop® Seal's properties include a 0,2 µm sterile barrier with a long-term resealable membrane for use with Ultra Yield™ and other brand flasks. Testing has been conducted on multiple customer samples with great results on up to seven days of cellular growth. TestingMore Product Information