Cleanroom wipes, TexTra™
Supplier: Texwipe
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TX2424
TEXWTX2424EA 789 GBP
TEXWTX2424
Cleanroom wipes, TexTra™
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Absorbent, single ply, 100% continuous filament polyester wipes, feature enhanced sorbency.
  • Economical, heavyweight, highly sorbent wipe, designed for non abrasive applications
  • Strong and durable fabric avoids snags, with good solvent, cleaning solution and disinfectant compatibility
  • Absorbency and liquid holding capacity of a two ply wipe, enables these wipes to be used in place of two ply wipes
Recommended for use with cleaning and disinfectant solutions, such as bleaches, phenols and quaternary ammonium compounds. Ideal for vigorous wiping,More Product Information