Racks for chest freezers, Alu FlexRack
Supplier: TENAK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TE20110 TE20112 TE20114 TE20116 TE20118 TE20120 TE20122 TE20124
TENATE20110EA 167 GBP
TENATE20110 TENATE20112 TENATE20114 TENATE20116 TENATE20118 TENATE20120 TENATE20122 TENATE20124
Racks for chest freezers, Alu FlexRack
Racks Cryogenic Box Racks
Aluminium.
  • Adjustable shelves - the rack can be used for all or a mixture of cryobox heights
  • Can be used for any chest freezer
  • Only buy the additional shelves you need
The Alu FlexRack is recommended for all applications where a flexible storage solution is needed.
Delivery information: Supplied with locking rod for securing boxes and one shelf.