Wash troughs
Supplier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
WTN010EX WTN010EXBAT WTN010EXBATPED WTN010EXPED WTN010EXSEN WTN010EXSENPED WTN020EX WTN020EXBAT WTN020EXBATPED WTN020EXPED WTN020EXSEN WTN020EXSENPED WTN030EX WTN030EXBAT WTN030EXPED WTN030EXSEN WTN030EXSENPED WTN040EX WTN040EXBATPED WTN040EXPED WTN040EXSEN WTN050EX WTN050EXBAT WTN050EXPED WTN050EXSEN WTN060EX WTN060EXBAT WTN060EXPED WTN060EXSEN WTN060EXSENPED
TEKMWTN010EXEA 598 GBP
TEKMWTN010EX TEKMWTN010EXBAT TEKMWTN010EXBATPED TEKMWTN010EXPED TEKMWTN010EXSEN TEKMWTN010EXSENPED TEKMWTN020EX TEKMWTN020EXBAT TEKMWTN020EXBATPED TEKMWTN020EXPED TEKMWTN020EXSEN TEKMWTN020EXSENPED TEKMWTN030EX TEKMWTN030EXBAT TEKMWTN030EXPED TEKMWTN030EXSEN TEKMWTN030EXSENPED TEKMWTN040EX TEKMWTN040EXBATPED TEKMWTN040EXPED TEKMWTN040EXSEN TEKMWTN050EX TEKMWTN050EXBAT TEKMWTN050EXPED TEKMWTN050EXSEN TEKMWTN060EX TEKMWTN060EXBAT TEKMWTN060EXPED TEKMWTN060EXSEN TEKMWTN060EXSENPED
Wash troughs
Faucets and Sinks Sinks
Our range of sensor operated wash troughs are manufactured from 304 grade stainless steel with a fully welded design for maximum hygiene. Choose from knee, battery or mains operation to suit your needs.
  • Secret wall fixing to hang the wash trough to to the wall
  • Supplied fitted, ready to plumb in
  • Infra-red sensor operated
  • Sloped trough for fast draining
Our wash troughs are manufactured in fully welded 304-grade stainless steel and can be wall-mounted or with additional pedestal support.
Being hands-free, the design supports rigorous hygienicMore Product Information