Front access PPE dispensers
Supplier: TEKNOMEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HWDP-F1V25X30 HWDP-F2V50X30 HWDP-F3V75X30 HWDP-F4V10X30 HWDP-F2V50X60 HWDP-F3V75X60 HWDP-F4V10X60 HWDPF1V25X30PE HWDPF2V50X30PE HWDPF3V75X30PE HWDPF4V10X30PE HWDPF1V25X60PE HWDPF2V50X60PE HWDPF3V75X60PE HWDPF4V10X60PE HWDI-FS5SD HWDI-WMFA25X60 HWDI-WMFA75X30 HWDI-WMFA50X30
TEKMHWDP-F1V25X30EA 337 GBP
TEKMHWDP-F1V25X30 TEKMHWDP-F2V50X30 TEKMHWDP-F3V75X30 TEKMHWDP-F4V10X30 TEKMHWDP-F2V50X60 TEKMHWDP-F3V75X60 TEKMHWDP-F4V10X60 TEKMHWDPF1V25X30PE TEKMHWDPF2V50X30PE TEKMHWDPF3V75X30PE TEKMHWDPF4V10X30PE TEKMHWDPF1V25X60PE TEKMHWDPF2V50X60PE TEKMHWDPF3V75X60PE TEKMHWDPF4V10X60PE TEKMHWDI-FS5SD TEKMHWDI-WMFA25X60 TEKMHWDI-WMFA75X30 TEKMHWDI-WMFA50X30
Front access PPE dispensers
PPE Dispensers
Front access PPE dispenser comes in a variety of sizes to suit most applications.
  • Brushed dull polish finish
  • No plastic parts
  • Mounts proud of wall or pedestal for easy cleaning
  • Pedestal fixings included
  • Fully welded material
The front access PPE dispenser is suitable for various items of PPE including beard snoods, hair nets, face masks, ear plugs and shoe covers. It has an ergonomic design with a wide dispensing tray and domed dispensing slot for easy access PPE to PPE.
Lifting tabs on the lid for ease of opening and filling, they double as aMore Product Information