ɑ-Methylene-γ-valerolactone ≥97.0% (by GC) stabilised

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 4,5-Dihydro-5-methyl-3-methylene-2(3H)-furanone

Stabilisation: Stabilised with hydroquinone (HQ)

M1453-25G
TCIAM1453-25GEA 0 GBP
TCIAM1453-25G
ɑ-Methylene-γ-valerolactone ≥97.0% (by GC) stabilised
alpha-Methylene-gamma-valerolactone
Formula: C₆H₈O₂
MW: 112.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06797118
CAS Number: 62873-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4e926c75aac8a915077f917ea79c32e7f534d25&vid=a4e926c75aac8a915077f917ea79c32e7f534d25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR