1-Chloro-2-methylbutane ≥95.0% (by GC)

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

C1459-5G
TCIAC1459-5GEA 0 GBP
TCIAC1459-5G
1-Chloro-2-methylbutane ≥95.0% (by GC)
1-Chloro-2-methylbutane
Formula: C₅H₁₁Cl
MW: 106.6 g/mol
Boiling Pt: 99°C
Flash Pt: 0°C
MDL Number: MFCD00039366
CAS Number: 616-13-7
EINECS: 210-466-9
UN: 1107
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e5673d933fa22992ec4c34e9dbe9768a1270e50&vid=0e5673d933fa22992ec4c34e9dbe9768a1270e50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR