SPE Plates, 96-well, NBE MAESTRO
Supplier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
613-0051NBE
TCAT613-0051NBEEA 0 GBP
TCAT613-0051NBE
SPE Plates, 96-well, NBE MAESTRO
Microplates Solid Phase Extraction Plates (SPE)
Silica-based sorbent with C18 bonding in proprietary coverage.
  • Reversed-phased
  • Liquid handler compatible
Uses a combination of filtration and sorbent interaction. Ideal for pass-through sample preparation techniques to remove phospholipids, proteins and particulates from sample matrices.