Cerex® TRACE-N20 SPE Column
Supplier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
611-0101R
TCAT611-0101REA 0 GBP
TCAT611-0101R
Cerex® TRACE-N20 SPE Column
Chromatography Columns
An end-capped C18 silica sorbent ideal for reverse-phase retention of neutral and polar analytes.
  • Reversed-phase
  • Liquid handler compatible
A 15 µm particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Formulated for 25-OH-VitD/steroid assays.