Atlas® NBE SCX SPE Plates, 96-well
Supplier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30174724 30174732
TCAT30174724EA 0 GBP
TCAT30174724 TCAT30174732
Atlas® NBE SCX SPE Plates, 96-well
Microplates Solid Phase Extraction Plates (SPE)
A hydrophilic/lipophilic balanced strong cationic chemistry offering excellent selectivity to basic and neutral compounds. Also utilising Tecan's unique NBE column platform. A direct alternative to Waters Oasis® MCX and Phenomenex Strata XC.
  • Base resin chemistry: N-vinylpyrrolidonedivinylbenzene copolymer
  • Surface chemistry: sulfonic acid
  • Particle packing: monodisperse porous microsphere
  • pH stability: 1 - 14
  • Simplified sample prep. protocol
  • Slurry packing ensures highest of quality, consistency and reproducibility
  • Available with SCX chemistry
  • NBE format
More Product Information